Skip to Content
Merck
All Photos(1)

Key Documents

591009

Sigma-Aldrich

2-Propylacrylic acid

99%

Synonym(s):

α-Propylacrylic acid, 2-Methylenepentanoic acid, Propacrylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,370.00

$1,370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,370.00

About This Item

Linear Formula:
CH3(CH2)2(=CH2)CO2H
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$1,370.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

contains

150 ppm BHT as inhibitor

refractive index

n20/D 1.441 (lit.)

bp

165-188 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC(=C)C(O)=O

InChI

1S/C6H10O2/c1-3-4-5(2)6(7)8/h2-4H2,1H3,(H,7,8)

InChI key

HEBDGRTWECSNNT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle M Hocking et al.
Tissue engineering. Part A, 25(5-6), 416-426 (2018-08-23)
Subarachnoid hemorrhage (SAH) is associated with vasospasm that is refractory to traditional vasodilators, and inhibition of vasospasm after SAH remains a large unmet clinical need. SAH causes changes in the phosphorylation state of the small heat shock proteins (HSPs), HSP20
Feng Qiu et al.
Biomaterials, 182, 82-91 (2018-08-15)
Cancer vaccines targeting patient-specific tumor neoantigens have recently emerged as a promising component of the rapidly expanding immunotherapeutic armamentarium. However, neoantigenic peptides typically elicit weak CD8+ T cell responses, and so there is a need for universally applicable vaccine delivery
Terence Ta et al.
Journal of controlled release : official journal of the Controlled Release Society, 194, 71-81 (2014-08-26)
Thermosensitive liposomes have emerged as a viable strategy for localized delivery and triggered release of chemotherapy. MR-guided focused ultrasound (MRgFUS) has the capability of heating tumors in a controlled manner, and when combined with thermosensitive liposomes can potentially reduce tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service