About This Item
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.5900 (lit.)
bp
141-142 °C (lit.)
density
1.189 g/mL at 25 °C (lit.)
functional group
chloro
phenoxy
SMILES string
ClCc1cccc(Oc2ccccc2)c1
InChI
1S/C13H11ClO/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-9H,10H2
InChI key
QUYVTGFWFHQVRO-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service