Skip to Content
Merck
All Photos(1)

Key Documents

588733

Sigma-Aldrich

(1R,2S,5R)-(−)-Menthol

≥99%, sublimed

Synonym(s):

(−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902293
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥99%

form

solid

optical activity

[α]20/D −50°, c = 10% in ethanol

bp

212 °C (lit.)

mp

41-45 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Voets et al.
Nature chemical biology, 3(3), 174-182 (2007-02-13)
TRPM8, a member of the transient receptor potential (TRP) channel superfamily, is expressed in thermosensitive neurons, in which it functions as a cold and menthol sensor. TRPM8 and most other temperature-sensitive TRP channels (thermoTRPs) are voltage gated; temperature and ligands
A Gelal et al.
Clinical pharmacology and therapeutics, 66(2), 128-135 (1999-08-25)
Menthol is widely used in a variety of commercial products and foods, but its clinical pharmacology is not well studied. To determine the disposition kinetics and to examine subjective and cardiovascular effects of menthol, we conducted a crossover placebo-controlled study
Tibor Rohács et al.
Nature neuroscience, 8(5), 626-634 (2005-04-27)
The subjective feeling of cold is mediated by the activation of TRPM8 channels in thermoreceptive neurons by cold or by cooling agents such as menthol. Here, we demonstrate a central role for phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)) in the activation of recombinant
Anne-Mette Haahr et al.
Physiology & behavior, 82(2-3), 531-540 (2004-07-28)
During chewing, the oral cavity functions like a bellow, forcing volatile flavour compounds into the exhaling air to the nasal compartment. Accordingly, we hypothesised that flavour release from chewing gum is predominantly governed by chewing frequency (CF), although other oral
Robert J Stein et al.
The Journal of urology, 172(3), 1175-1178 (2004-08-18)
Overactive bladder symptoms due to various etiologies have been successfully treated with capsaicin by desensitization of the temperature sensitive vanilloid receptor TRPV1. Recently another temperature sensitive receptor, TRPM8, activated by menthol and cool temperatures (8C to 28C) was described that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service