Skip to Content
Merck
All Photos(1)

Key Documents

58870

Sigma-Aldrich

(−)-Isoledene

≥95.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$615.00

$615.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$615.00

About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2085196
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$615.00


Estimated to ship on02 June 2025


Request a Bulk Order

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D −50.5±1°, neat

refractive index

n20/D 1.493

bp

95 °C/5 mmHg (lit.)

density

0.902 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@H]2[C@H](C3=C1CC[C@H]3C)C2(C)C

InChI

1S/C15H24/c1-9-6-8-12-14(15(12,3)4)13-10(2)5-7-11(9)13/h9-10,12,14H,5-8H2,1-4H3/t9-,10-,12-,14-/m1/s1

InChI key

NUQDPKOFUKFKFD-BGOOENEXSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service