Skip to Content
Merck
All Photos(1)

Key Documents

579653

Sigma-Aldrich

Naphthalene-13C10

99 atom % 13C

Synonym(s):

Naphthalene-ul-13C

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$4,230.00

$4,230.00


Check Cart for Availability


Select a Size

Change View
50 MG
$4,230.00

About This Item

Linear Formula:
13C10H8
CAS Number:
Molecular Weight:
138.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

$4,230.00


Check Cart for Availability

isotopic purity

99 atom % 13C

Quality Level

bp

218 °C (lit.)

mp

80-82 °C (lit.)

application(s)

environmental

mass shift

M+10

SMILES string

[13cH]1[13cH][13cH][13c]2[13cH][13cH][13cH][13cH][13c]2[13cH]1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1

InChI key

UFWIBTONFRDIAS-IIYFYTTLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service