Skip to Content
Merck
All Photos(2)

Documents

579068

Sigma-Aldrich

Potassium 4-methoxyphenyltrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4BF3K
CAS Number:
Molecular Weight:
214.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

>300 °C (lit.)

SMILES string

[K+].COc1ccc(cc1)[B-](F)(F)F

InChI

1S/C7H7BF3O.K/c1-12-7-4-2-6(3-5-7)8(9,10)11;/h2-5H,1H3;/q-1;+1

InChI key

XNYMCUFKZHRYID-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Potassium trifluoroborates are moisture- and air-stable organoboron reagents suitable for oxidative conditions.

Related Content

The central theme of the Molander group's research is the development of new synthetic methods and their application to the synthesis of organic molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service