578452
1-Pentenylboronic acid
≥95%
Synonym(s):
1-Penten-1-ylboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H11BO2
CAS Number:
Molecular Weight:
113.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
solid
mp
100-105 °C (lit.)
SMILES string
[H]\C(CCC)=C(\[H])B(O)O
InChI
1S/C5H11BO2/c1-2-3-4-5-6(7)8/h4-5,7-8H,2-3H2,1H3/b5-4+
InChI key
SYKWJOZHNDPWIM-SNAWJCMRSA-N
Application
Reactant for:
- Synthesis of 8-alkenylborondipyrromethene dyes via Liebeskind-Srogl cross-coupling starting from 8-thiomethyl-substituted BODIPY
- Rhodium and palladium catalyzed substitution reactions with enyne acetates and carbonates
- Halogenation and protodeauration reactions of organogold phosphine complexes
- Lewis acid catalyzed Nazarov reaction
- Palladium-catalyzed tandem intramolecular C-N and intermolecular Suzuki coupling process
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service