All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
248-252 °C (lit.)
SMILES string
Oc1ccc2c(OC(=O)C=C2c3ccccc3)c1
InChI
1S/C15H10O3/c16-11-6-7-12-13(10-4-2-1-3-5-10)9-15(17)18-14(12)8-11/h1-9,16H
InChI key
IVJMJRRORVMRJJ-UHFFFAOYSA-N
Gene Information
mouse ... Maoa(17161)
rat ... Aldh1a2(116676)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service