Skip to Content
Merck
All Photos(1)

Key Documents

573019

Sigma-Aldrich

Ethylene carbonate-d4

98 atom % D, 99% (CP)

Synonym(s):

1,3-Dioxolan-2-one-d4, Deuterated ethylene carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,420.00
5 G
$4,660.00

$1,420.00


Check Cart for Availability


Select a Size

Change View
1 G
$1,420.00
5 G
$4,660.00

About This Item

Empirical Formula (Hill Notation):
C3D4O3
CAS Number:
Molecular Weight:
92.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.13

$1,420.00


Check Cart for Availability

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

246 °C

mp

34-37 °C

density

1.379 g/mL at 25 °C

mass shift

M+4

storage temp.

room temp

SMILES string

[2H]C1([2H])OC(=O)OC1([2H])[2H]

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2/i1D2,2D2

InChI key

KMTRUDSVKNLOMY-LNLMKGTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katie L Browning et al.
Physical chemistry chemical physics : PCCP, 21(31), 17356-17365 (2019-07-30)
With the use of in situ neutron reflectometry (NR) we show how the addition of an electronically conductive polymeric binder, PEFM, mediates the solid-electrolyte interphase (SEI) formation and composition on an amorphous Si (a-Si) electrode as a function of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service