Skip to Content
Merck
All Photos(2)

Key Documents

570249

Sigma-Aldrich

Fluorescein O,O′-dimethacrylate

95%

Synonym(s):

3’,6′-Dimethacryloxyspirobenzo[c]-furan[1,9’]xanthen-3-one, Dimethacryloyloxyfluorescein

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$392.00

$392.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$392.00

About This Item

Empirical Formula (Hill Notation):
C28H20O7
CAS Number:
Molecular Weight:
468.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$392.00


Available to ship on31 March 2025Details


Request a Bulk Order

Assay

95%

form

solid

mp

154.5-156.0 °C (lit.)

λmax

203 nm

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Oc1ccc2c(Oc3cc(OC(=O)C(C)=C)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C28H20O7/c1-15(2)25(29)32-17-9-11-21-23(13-17)34-24-14-18(33-26(30)16(3)4)10-12-22(24)28(21)20-8-6-5-7-19(20)27(31)35-28/h5-14H,1,3H2,2,4H3

InChI key

IJMJZIOPZNRQMM-UHFFFAOYSA-N

Application

Fluorescent monomer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service