Skip to Content
Merck
All Photos(1)

Key Documents

566780

Sigma-Aldrich

2-Acetylanthracene

98%

Synonym(s):

1-(2-Anthracenyl)acetone, 1-(2-Anthracenyl)ethanone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$174.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$174.00

About This Item

Empirical Formula (Hill Notation):
C16H12O
CAS Number:
Molecular Weight:
220.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$174.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

189-192 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccc2cc3ccccc3cc2c1

InChI

1S/C16H12O/c1-11(17)12-6-7-15-9-13-4-2-3-5-14(13)10-16(15)8-12/h2-10H,1H3

InChI key

DQFWGPPCMONVDS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kanji Nagai et al.
Journal of chromatography. A, 1549, 85-92 (2018-03-28)
Poly(butylene terephthalate) based novel stationary phase (SP), composed of planar aromatic phenyl group together with ester group monomer units, was designed for supercritical fluid chromatography (SFC) use. As expected from its structure, this phase shows planarity recognition of isomeric aromatics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service