Skip to Content
Merck
All Photos(1)

Documents

566136

Sigma-Aldrich

Dimethylbis(pentafluorophenyl)tin

Synonym(s):

Bis(pentafluorophenyl)dimethylstannane, Bis(pentafluorophenyl)dimethyltin, Dimethylbis(pentafluorophenyl)stannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6F5)2Sn(CH3)2
CAS Number:
Molecular Weight:
482.89
MDL number:
UNSPSC Code:
12352300

refractive index

n20/D 1.4910 (lit.)

bp

259-260 °C (lit.)

density

1.827 g/mL at 25 °C (lit.)

Application

Mild reagent for the selective metathesis of B-Cl to B-C6F5 with little or no competing alkyl exchange.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service