Skip to Content
Merck
All Photos(1)

Documents

565040

Sigma-Aldrich

3-Chloro-4-methoxybenzaldehyde

97%

Synonym(s):

3-Chloro-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3ClCHO
CAS Number:
Molecular Weight:
170.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

56-60 °C (lit.)

SMILES string

[H]C(=O)c1ccc(OC)c(Cl)c1

InChI

1S/C8H7ClO2/c1-11-8-3-2-6(5-10)4-7(8)9/h2-5H,1H3

InChI key

WYVGYYIZXPXHAZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongfei Han et al.
PloS one, 8(9), e73350-e73350 (2013-09-26)
A novel flavoprotein monooxygenase, trans-anethole oxygenase (TAO), from Pseudomonas putida JYR-1, which is capable of catalyzing the oxidation of trans-anethole to p-anisaldehyde, was heterologously expressed in E. coli and purified. Enzymatic kinetics of diverse substrates and cofactors revealed that TAO
E de Jong et al.
FEBS letters, 305(3), 220-224 (1992-07-06)
The white-rot fungus Bjerkandera sp. BOS55 produced de-novo several aromatic metabolites. Besides veratryl alcohol and veratraldehyde, compounds which are known to be involved in the ligninolytic system of several other white-rot fungi, other metabolites were formed. These included anisaldehyde, 3-chloro-anisaldehyde
T Mester et al.
Applied and environmental microbiology, 63(5), 1987-1994 (1997-05-01)
Aryl metabolites are known to have an important role in the ligninolytic system of white rot fungi. The addition of known precursors and aromatic acids representing lignin degradation products stimulated the production of aryl metabolites (veratryl alcohol, veratraldehyde, p-anisaldehyde, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service