Skip to Content
Merck
All Photos(2)

Key Documents

562939

Sigma-Aldrich

2-Amino-4-chlorophenyl phenyl ether

97%

Synonym(s):

5-Chloro-2-phenoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)OC6H5
CAS Number:
Molecular Weight:
219.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

41-44 °C (lit.)

SMILES string

Nc1cc(Cl)ccc1Oc2ccccc2

InChI

1S/C12H10ClNO/c13-9-6-7-12(11(14)8-9)15-10-4-2-1-3-5-10/h1-8H,14H2

InChI key

SXEBHIMOUHBBOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service