Skip to Content
Merck
All Photos(2)

Key Documents

560162

Sigma-Aldrich

3-Bromo-2-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$181.00

$181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$181.00

About This Item

Linear Formula:
Br(CH3)C6H3COOH
CAS Number:
Molecular Weight:
215.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$181.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

152-156 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

Cc1c(Br)cccc1C(O)=O

InChI

1S/C8H7BrO2/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4H,1H3,(H,10,11)

InChI key

BJGKVCKGUBYULR-UHFFFAOYSA-N

Application

3-Bromo-2-methylbenzoic acid may be used to synthesize 4-(bromoacetyl)isoindolin-1-one[1] and isocoumarins.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New anti-viral drugs for the treatment of the common cold.
Maugeri C, et al.
Bioorganic & Medicinal Chemistry, 16(6), 3091-3107 (2008)
Mapping the ATP-binding domain of DNA-dependent protein kinase (DNA-PK) with coumarin-and isocoumarin-derived inhibitors.
Payne SL, et al.
Bioorganic & Medicinal Chemistry Letters, 20(12), 3649-3653 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service