560081
9,9-Di-(2′-ethylhexyl)-2,7-dibromofluorene
98%
Synonym(s):
2,7-Dibromo-9,9-bis(2-ethylhexyl)-9H -fluorene, 2,7-Dibromo-9,9-bis(2-ethylhexyl)fluorene, 2,7-Dibromo-9,9-di-2-ethylhexylfluorene, 9,9-Bis(2-ethylhexyl)-2,7-dibromofluorene
About This Item
Recommended Products
Assay
98%
refractive index
n20/D 1.5720 (lit.)
density
1.230 g/mL at 25 °C (lit.)
SMILES string
CCCCC(CC)CC1(CC(CC)CCCC)c2cc(Br)ccc2-c3ccc(Br)cc13
InChI
1S/C29H40Br2/c1-5-9-11-21(7-3)19-29(20-22(8-4)12-10-6-2)27-17-23(30)13-15-25(27)26-16-14-24(31)18-28(26)29/h13-18,21-22H,5-12,19-20H2,1-4H3
InChI key
WSBUZOZAKDVRTK-UHFFFAOYSA-N
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service