Skip to Content
Merck
All Photos(1)

Key Documents

55787

Sigma-Aldrich

(±)-3-(Boc-aminomethyl)piperidine

≥98.0% (TLC)

Synonym(s):

tert-Butyl N-(3-piperidylmethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
Beilstein:
7023643
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCC1CCCNC1

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-8-9-5-4-6-12-7-9/h9,12H,4-8H2,1-3H3,(H,13,14)

InChI key

KHPQHXGYYXYTDN-UHFFFAOYSA-N

Application

Reagent for:
Preparation of Tylophorine analogs
Enzymatic acylation

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mono-Boc-protected diamines are versatile building blocks for chemical synthesis. Their production is a lot more challenging than the simple reaction scheme might imply, because the Boc-anhydride reagent cannot differentiate between the two identical amino moieties in the substrate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service