Skip to Content
Merck
All Photos(2)

Key Documents

557374

Sigma-Aldrich

Tris(acetonitrile)tricarbonylchromium(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(CO)3(NCCH3)3
CAS Number:
Molecular Weight:
259.18
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

form

solid

reaction suitability

core: chromium
reagent type: catalyst

mp

67-72 °C (dec.) (lit.)

SMILES string

[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC#N.CC#N.CC#N

InChI

1S/3C2H3N.3CO.Cr/c3*1-2-3;3*1-2;/h3*1H3;;;;

InChI key

XPEKEFWOCFTTML-UHFFFAOYSA-N

Application

  • Catalyst for thermal transformation of poly(methylsilene) to SiC ceramics via crosslinking reactions
  • Catalyst for reduction of nitrobenzenes and nitrosobenzenes

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service