Skip to Content
Merck
All Photos(1)

Key Documents

557323

Sigma-Aldrich

o-Toluenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2Cl
CAS Number:
Molecular Weight:
190.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D >1.5580 (lit.)

bp

254 °C (lit.)

density

1.320 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1S(Cl)(=O)=O

InChI

1S/C7H7ClO2S/c1-6-4-2-3-5-7(6)11(8,9)10/h2-5H,1H3

InChI key

HDECRAPHCDXMIJ-UHFFFAOYSA-N

General description

o-Toluenesulfonyl chloride can be synthesized by reacting o-thiocresol in glacial acetic acid with chlorine.[1] It participates in the preparation of 5-chloro-3-phenyl-2,1-benzisoxazole.[2]

Application

o-Toluenesulfonyl chloride may be used in the synthesis of:
  • oxazoline[3]
  • mesityl o-tolyl sulfone[1]
  • allyl o-toluenesulfonate[4]
  • 2-methyl-4′-t-butyldiphenyl sulfone[5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intramolecular nucleophilic participation. V. The role of the ortho-substituent in the solvolysis of o-nitrobenzhydrl bromide and o-nitrobenzyl tosylate.
Mease AD, et al.
Journal of the American Chemical Society, 90(7), 1797-1801 (1968)
The Metalation of 4-t-Butyldiphenyl Sulfone with n-Butyllithium1.
Shirley DA and Lehto EA.
Journal of the American Chemical Society, 79(13), 3481-3485 (1957)
Min Wang et al.
Biomaterials, 261, 120301-120301 (2020-09-02)
Local tumor therapy through injectable biodegradable hydrogels with controlled drug release has attracted much attention recently, due to their easy operation, low side effect and efficiency. However, most of the reported therapeutic hydrogel system showed a lack of biodegradation tracking
Cyclization of Acylaminoalkanols to 2-Oxazolines1.
Boyd RN and Rittner RC.
Journal of the American Chemical Society, 82(8), 2032-2034 (1960)
Rearrangements of sulfones to sulfinic acids via carbanion intermediates.
Truce WE and Brand WW
The Journal of Organic Chemistry, 35(6), 1828-1833 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service