Skip to Content
Merck
All Photos(1)

Key Documents

556394

Sigma-Aldrich

Tris[4-(diethylamino)phenyl]amine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$42.80

$42.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$42.80

About This Item

Linear Formula:
[(C2H5)2NC6H4]3N
CAS Number:
Molecular Weight:
458.68
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$42.80


Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

104-106 °C (lit.)

SMILES string

CCN(CC)c1ccc(cc1)N(c2ccc(cc2)N(CC)CC)c3ccc(cc3)N(CC)CC

InChI

1S/C30H42N4/c1-7-31(8-2)25-13-19-28(20-14-25)34(29-21-15-26(16-22-29)32(9-3)10-4)30-23-17-27(18-24-30)33(11-5)12-6/h13-24H,7-12H2,1-6H3

InChI key

HWNGZPYALGWORF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate for photochromic dye. Hole transporting material.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmad Arar et al.
Polymers, 13(2) (2021-01-23)
Redox initiating systems (RISs) are highly worthwhile for polymerization in mild conditions (at room temperature-RT) without external thermal or light activation. With high performance redox initiating systems RIS, the free radical polymerization FRP can even be carried out under air

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service