Skip to Content
Merck
All Photos(3)

Key Documents

556157

Sigma-Aldrich

2-Cyanobenzenesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4SO2Cl
CAS Number:
Molecular Weight:
201.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

65-69 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccccc1C#N

InChI

1S/C7H4ClNO2S/c8-12(10,11)7-4-2-1-3-6(7)5-9/h1-4H

InChI key

NQAYCMBZPAARNO-UHFFFAOYSA-N

Application

2-Cyanobenzenesulfonyl chloride may be used in the synthesis of:
  • 2-butyl-5-(2-cyanophenyl)furan[1]
  • 2-(2-cyanophenyl)-3,6-dimethyl-4,5,6,7-tetrahydrobenzofuran[1]
  • 5-thia-4b,10-diaza-indeno[2,1-a]indene-5,5-dioxide[2]
  • 1-methyl-2-(2-cyanophenyl)pyrrole[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-Assisted, One-Pot Synthesis of 5-Thia-4 b, 10-diaza-indeno [2, 1-a]-indene-5, 5-dioxide and 10 H-11-Thia-5, 10 a-diaza-benzo [b] fluorene-11, 11-dioxide.
Cole AG, et al.
Synthetic Communications, 39(20), 3607-3610 (2009)
Palladium-Catalysed Direct Desulfitative Arylation of Pyrroles using Benzenesulfonyl Chlorides as Alternative Coupling Partners.
Jin R, et al.
Advanced Synthesis & Catalysis, 356(18), 3831-3841 (2014)
Benzenesulfonyl Chlorides: Alternative Coupling Partners for Regiocontrolled Palladium-Catalyzed Direct Desulfitative 5-Arylation of Furans.
Beladhria A, et al.
Synthesis, 46(18), 2515-2523 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service