Skip to Content
Merck
All Photos(1)

Key Documents

555886

Sigma-Aldrich

3-((tert-Butyldimethylsilyl)oxy)-propanol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$119.00
25 ML
$563.00

$119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$119.00
25 ML
$563.00

About This Item

Linear Formula:
(CH3)3CSi(CH3)2OCH2CH2CH2OH
CAS Number:
Molecular Weight:
190.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.4350 (lit.)

bp

110 °C/6 mmHg (lit.)

density

0.892 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)[Si](C)(C)OCCCO

InChI

1S/C9H22O2Si/c1-9(2,3)12(4,5)11-8-6-7-10/h10H,6-8H2,1-5H3

InChI key

NETUFVYVNJNFMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-((tert-Butyldimethylsilyl)oxy)-propanol (3-[(tert-butyldimethylsilyl)oxy]-1-propanol) is formed as an intermediate during the synthesis of 3-(tert-butyldimethylsilyl)oxy]propanal.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.0 °F - closed cup

Flash Point(C)

92.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective epoxidation of Phe-Gly and Phe-Phe vinyl isosteresStereoselective epoxidation of Phe-Gly and Phe-Phe vinyl isosteres.
Jenmalm A, et al.
The Journal of Organic Chemistry, 59(5), 1139-1148 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service