Skip to Content
Merck
All Photos(2)

Key Documents

552887

Sigma-Aldrich

3-(Trimethylammonium)propyl-functionalized silica gel, carbonate

200-400 mesh, extent of labeling: 0.8 mmol/g loading

Synonym(s):

TMA carbonate-3 functionalized silica gel

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$293.00

$293.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$293.00

About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$293.00


Check Cart for Availability

Request a Bulk Order

extent of labeling

0.8 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

SMILES string

O=[SiH2].O=[SiH2].[O-]C([O-])=O.CCC[N+](C)(C)C.CCC[N+](C)(C)C

InChI

1S/2C6H16N.CH2O3.2H2OSi/c2*1-5-6-7(2,3)4;2-1(3)4;2*1-2/h2*5-6H2,1-4H3;(H2,2,3,4);2*2H2/q2*+1;;;/p-2

InChI key

WMFSAHWNSGLABT-UHFFFAOYSA-L

Application

Silica-bound base and acid scavenger including acidic phenols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service