Skip to Content
Merck
All Photos(1)

Key Documents

55100

Sigma-Aldrich

8-Quinolinol hemisulfate salt

≥98.0% (dry substance, T), yellow

Synonym(s):

8-Hydroxyquinoline hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O2 · H2SO4
CAS Number:
Molecular Weight:
388.39
Beilstein:
3760550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

C9H7NO · ½H2SO4 (Linear Formula)

Assay

≥98.0% (dry substance, T)

impurities

~5% water

color

yellow

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

OS(O)(=O)=O.Oc1cccc2cccnc12.Oc3cccc4cccnc34

InChI

1S/2C9H7NO.H2O4S/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4/h2*1-6,11H;(H2,1,2,3,4)

InChI key

YYVFXSYQSOZCOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

8-Quinolinol hemisulfate salt (8-OHQ) may be used in the preparation of Cu[8-OHQ]2, which shows proteasome-inhibitory activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenyon G Daniel et al.
Biochemical pharmacology, 67(6), 1139-1151 (2004-03-10)
Here we report that organic copper complexes can potently and selectively inhibit the chymotrypsin-like activity of the proteasome in vitro and in vivo. Several copper compounds, such as NCI-109268 and bis-8-hydroxyquinoline copper(II) [Cu(8-OHQ)(2)], can inhibit the chymotrypsin-like activity of purified
Erik H J G Aarntzen et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(6), 1525-1533 (2013-02-06)
Anticancer dendritic cell (DC) vaccines require the DCs to relocate to lymph nodes (LN) to trigger immune responses. However, these migration rates are typically very poor. Improving the targeting of ex vivo generated DCs to LNs might increase vaccine efficacy
Saverio Tardito et al.
Journal of medicinal chemistry, 55(23), 10448-10459 (2012-11-23)
This study reports the structure-activity relationship of a series of 8-hydroxoquinoline derivatives (8-HQs) and focuses on the cytotoxic activity of 5-Cl-7-I-8-HQ (clioquinol, CQ) copper complex (Cu(CQ)). 8-HQs alone cause a dose-dependent loss of viability of the human tumor HeLa and
Meng Zhang et al.
Inorganic chemistry, 51(21), 11315-11323 (2012-10-09)
An artificial peptide with three pendant hydroxyquinoline (hq) ligands on a palindromic backbone was designed and used to form multimetallic assemblies. Reaction of the tripeptide with zinc acetate led to a highly fluorescent tripeptide duplex with three Zn(II) coordinative cross-links.
Bhimashankar Mitkari et al.
Experimental neurology, 239, 158-162 (2012-10-13)
Cell therapies from various sources have been under intense research in stroke. Efficient homing of the cells to the injured brain without complications is necessary to realize the therapeutic potential of cell therapy. Intra-arterial (IA) infusion of cells bypasses the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service