Skip to Content
Merck
All Photos(1)

Key Documents

550817

Sigma-Aldrich

2-(1,3,3-Trimethylindolin-2-ylidene)acetaldehyde

97%

Synonym(s):

(1,3,3-Trimethylindolin-2-ylidene)acetaldehyde, 1,3,3-Trimethyl-Δ2,α-indolineacetaldehyde, 1,3,3-Trimethyl-2-(formylmethylene)-2,3-dihydroindole, 1,3,3-Trimethyl-2-methyleneindoline-ω-aldehyde, Fischer′s aldehyde, NSC 68048

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$378.00

$378.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$378.00

About This Item

Empirical Formula (Hill Notation):
C13H15NO
CAS Number:
Molecular Weight:
201.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$378.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

112-116 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

CN1c2ccccc2C(C)(C)\C1=C/C=O

InChI

1S/C13H15NO/c1-13(2)10-6-4-5-7-11(10)14(3)12(13)8-9-15/h4-9H,1-3H3/b12-8+

InChI key

GCECACVNILMTRD-XYOKQWHBSA-N

General description

2-(1,3,3-Trimethylindolin-2-ylidene)acetaldehyde is an a,ß-unsaturated aldehyde.

Application

2-(1,3,3-Trimethylindolin-2-ylidene)acetaldehyde may be used in the preparation of 1,3,3-trimethyl-2-anilino-vinyl-3-H-indolium salts, which are mainly used as hemicyanine dyes in textile industry.[1]
Reactant for preparation of:
  • Cyanine dyes
  • Photopolymer holographic recording materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterisation of 3-H-indolo-2-dimethinehemicyanine dyes obtained by the condensation of (1,3,3-trimethylindolin-2-ylidene) acetaldehyde with anilines in acetic acid.
Gaspar CL, et al.
Dyes and Pigments, 74(2), 306-312 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service