Skip to Content
Merck
All Photos(1)

Key Documents

549940

Sigma-Aldrich

6-Chlorohexanoyl chloride

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$586.00

$586.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$586.00

About This Item

Linear Formula:
Cl(CH2)5COCl
CAS Number:
Molecular Weight:
169.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$586.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.4640 (lit.)

density

1.400 g/mL at 25 °C (lit.)

functional group

acyl chloride
chloro

SMILES string

ClCCCCCC(Cl)=O

InChI

1S/C6H10Cl2O/c7-5-3-1-2-4-6(8)9/h1-5H2

InChI key

WZILXAPNPKMOSA-UHFFFAOYSA-N

Application

6-Chlorohexanoyl chloride may be used in the following processes:
  • Preparation of tert-butyl-N-(2-(6-chlorovaleryl)phenyl)carbamate, an intermediate for synthesizing the ketone analog of 2-amino-N-(4-(4-(1,2-benzisothiazol-3-yl)-1-piperazinyl)-butyl)benzamide hydrochloride.[1]
  • Preparation of (S)-(-)6-chlorohexanoic acid (2-methoxymethylpyrrolidin-1-yl)amide, a precursor for synthesizing 2-alkyl-substituted lactones.[2]
  • Deprotection and acylation of the pendant nitrogen during the preparation of a new cryptand with hydroxamate moiety that can bind with tribasic cations.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Enantioselective Synthesis of 2-Substituted 6-and 7-Membered Lactones via a-Alkylation of Lactone Hydrazones.
Enders D, et al.
Synthesis, 1995(08), 947-951 (1995)
Analogues of the potential antipsychotic agent 1192U90: amide modifications.
Navas III F, et al.
Bioorganic & Medicinal Chemistry, 6(6), 811-823 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service