Skip to Content
Merck
All Photos(1)

Key Documents

549126

Sigma-Aldrich

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5050 (lit.)

density

1.265 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1snnc1C

InChI

1S/C6H8N2O2S/c1-3-10-6(9)5-4(2)7-8-11-5/h3H2,1-2H3

InChI key

AHPXTXGCMLOXGA-UHFFFAOYSA-N

General description

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate is a bioactive nitrogen-containing heterocycle.[1]

Application

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate may be used in the preparation of following compounds with potent fungicidal activity:
  • 1,2,3-thiadiazole bearing hydrazone derivatives[2]
  • 2-(4′-methyl-1′,2′,3′-thiadiazol)-5-substituted-1,3,4-oxadiazole derivatives[3]
  • 1,2,3-thiadiazole bearing 1,2,4-triazole derivatives[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave assisted synthesis, antifungal activity and DFT theoretical study of some novel 1, 2, 4-triazole derivatives containing the 1, 2, 3-thiadiazole moiety.
Sun NB, et al.
Molecules (Basel), 18(10), 12725-12739 (2013)
Zhijin Fan et al.
Journal of agricultural and food chemistry, 57(10), 4279-4286 (2009-05-27)
Elicitors provide a broad spectrum of systemic acquired resistance by altering the physical and physiological status of the host plants and, therefore, are among the most successful directions in modern pesticide development for plant protection. To develop a novel elicitor
Synthesis of tetrazole containing 1, 2, 3-thiadiazole derivatives via U-4CR and their anti-TMV activity.
Wang SX, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 24(10), 889-892 (2013)
Microwave synthesis and biological activity of hydrazone derivatives containing 1, 2, 3-thiadiazole.
Liu XH, et al.
Asian Journal of Chemistry, 23(9), 4064-4064 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service