Skip to Content
Merck
All Photos(1)

Key Documents

549061

Sigma-Aldrich

3-(Dimethylamino)benzyl alcohol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$117.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$117.00

About This Item

Linear Formula:
(CH3)2NC6H4CH2OH
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.577 (lit.)

bp

282-283 °C (lit.)

density

1.056 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CN(C)c1cccc(CO)c1

InChI

1S/C9H13NO/c1-10(2)9-5-3-4-8(6-9)7-11/h3-6,11H,7H2,1-2H3

InChI key

YTUXZVSDDHCTBZ-UHFFFAOYSA-N

Application

3-(Dimethylamino)benzyl alcohol may be used in the preparation of (3-hydroxymethylphenyl)trimethylammonium iodide by reacting with methyl iodide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asse Marjasvaara et al.
Journal of mass spectrometry : JMS, 39(10), 1139-1146 (2004-10-07)
Substituted benzyl alcohol was oxidized enzymatically with a laccase-mediator system and the products were investigated as a function of time by nanoelectrospray ionization Fourier transform ion cyclotron resonance mass spectrometry (nanoESI-FTICRMS). With Trametes versicolor laccase (TVL), the mediator, 2,2',6,6'-tetramethylpiperidine-N-oxyl radical
Haifaa Mokbel et al.
Macromolecular rapid communications, 41(15), e2000289-e2000289 (2020-07-09)
Free radical polymerization upon near-infrared (NIR) light is still the subject of intense research efforts and remains a huge challenge particularly for long wavelengths (>1000 nm). In this study, a NIR sensitizer operating upon long wavelength (1064 nm) is proposed for an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service