Skip to Content
Merck
All Photos(1)

Documents

546542

Sigma-Aldrich

5-Chloro-2-nitrobenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4(NO2)CONH2
CAS Number:
Molecular Weight:
200.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

157-160 °C (lit.)

SMILES string

NC(=O)c1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C7H5ClN2O3/c8-4-1-2-6(10(12)13)5(3-4)7(9)11/h1-3H,(H2,9,11)

InChI key

MKHXTOPPKVFSFI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and preliminary cytotoxic and antifungal evaluation of some 6-N,N-dialkyl 2-aryl-4(3H)-quinazolinone derivatives.
Lopez SE, et al.
Heterocyclic Communications, 7(5), 473-480 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service