Skip to Content
Merck
All Photos(1)

Key Documents

545333

Sigma-Aldrich

1,2-Dichlorohexafluorocyclopentene

97%

Synonym(s):

1,2-Dichloro-3,3,4,4,5,5-hexafluoro-1-cyclopentene, 1,2-Dichloro-3,3,4,4,5,5-hexafluorocyclopentene, 1,2-Dichlorohexafluorocyclopent-1-ene, 1,2-Dichloroperfluorocyclopentene, 3,3,4,4,5,5-Hexafluoro-1,2-dichlorocyclopentene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$277.00

$277.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$277.00

About This Item

Linear Formula:
Cl2C5F6
CAS Number:
Molecular Weight:
244.95
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$277.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.37 (lit.)

bp

90 °C (lit.)

density

1.635 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

FC1(F)C(Cl)=C(Cl)C(F)(F)C1(F)F

InChI

1S/C5Cl2F6/c6-1-2(7)4(10,11)5(12,13)3(1,8)9

InChI key

ABPBVCKGWWGZDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichlorohexafluorocyclopentene is a halogenated cyclopentene derivative.

Application

1,2-Dichlorohexafluorocyclopentene may be used to synthesize:
  • inverse

  • diarylperfluorocyclopentenes[1]
  • fluoro-bisthienylcyclopentenes (F-BTCs)[2]
  • 1,1-dichlorooctafluorocyclopentane[3]
  • 1,2-dichlorooctafluorocyclopentane[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel strategy for synthesis of dichlorooctafluorocyclopentane and reaction mechanism investigation.
Zhang P, et al.
Journal of Fluorine Chemistry, 186, 33-39 (2016)
Perfluorocyclohexene bridges in inverse DiArylEthenes: synthesis through Pd-catalysed C?H bond activation, experimental and theoretical studies on their photoreactivity.
Yuan K, et al.
Chemical Communications (Cambridge, England), 49(72), 7896-7898 (2013)
Synthesis of Symmetrical and Nonsymmetrical Bisthienylcyclopentenes.
Szaloki G and Pozzo JL.
Chemistry (Weinheim An Der Bergstrasse, Germany), 19(34), 11124-11132 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service