Skip to Content
Merck
All Photos(1)

Documents

543748

Sigma-Aldrich

6-Fluoro-4-hydroxycoumarin

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5FO3
CAS Number:
Molecular Weight:
180.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

248-251 °C (lit.)

SMILES string

OC1=CC(=O)Oc2ccc(F)cc12

InChI

1S/C9H5FO3/c10-5-1-2-8-6(3-5)7(11)4-9(12)13-8/h1-4,11H

InChI key

JCOYNGQUPPGRNB-UHFFFAOYSA-N

General description

6-Fluoro-4-hydroxycoumarin undergoes direct sulfanylation with thiols in water to yield corresponding 4-sulfanylcoumarins.

Application

6-Fluoro-4-hydroxycoumarin may be used to synthesize chromenopyridinones.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eco-friendly construction of highly functionalized chromenopyridinones by an organocatalyzed solid-state melt reaction and their optical properties.
Paul S and Lee YR.
Green Chemistry, 18(6), 1488-1494 (2016)
Direct sulfanylation of 4-hydroxycoumarins with thiols in water.
Peng YY, et al.
Tetrahedron Letters, 50(20), 2405-2406 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service