Skip to Content
Merck
All Photos(1)

Key Documents

541036

Sigma-Aldrich

Silica, mesostructured

HMS (wormhole)

Synonym(s):

Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

surface area

800-1000 m2/g

pore size

1-2 cm3/g pore volume
2-4 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K E Kovalenko et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1048-1060 (2013-08-24)
Boreal wetlands play an important role in global carbon balance. However, their ecosystem function is threatened by direct anthropogenic disturbance and climate change. Oil sands surface mining in the boreal regions of Western Canada denudes tracts of land of organic
Fabrice Gritti et al.
Journal of chromatography. A, 1342, 24-29 (2014-04-17)
The advantages of using columns packed with stationary phases having a composition gradient so that retention factors increase toward the outlet and eluting them with an isocratic mobile phase may provide increased peak resolution in liquid chromatography. This approach is
C J Horwell et al.
Environmental research, 127, 63-73 (2013-11-26)
The six week eruption of Eyjafjallajökull volcano in 2010 produced heavy ash fall in a sparsely populated area of southern and south eastern Iceland and disrupted European commercial flights for at least 6 days. We adopted a protocol for the
Yang Ding et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10123-10128 (2013-06-05)
Undulatory locomotion, a gait in which thrust is produced in the opposite direction of a traveling wave of body bending, is a common mode of propulsion used by animals in fluids, on land, and even within sand. As such, it
Graham J Edgar et al.
Nature, 506(7487), 216-220 (2014-02-07)
In line with global targets agreed under the Convention on Biological Diversity, the number of marine protected areas (MPAs) is increasing rapidly, yet socio-economic benefits generated by MPAs remain difficult to predict and under debate. MPAs often fail to reach

Articles

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service