Skip to Content
Merck
All Photos(1)

Key Documents

540986

Sigma-Aldrich

1-Ethyl-2-nitrobenzene

96%

Synonym(s):

2-Nitroethylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$61.80

$61.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$61.80

About This Item

Linear Formula:
C2H5C6H4NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
1865655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.80


Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.537 (lit.)

bp

172-174 °C/18 mmHg (lit.)

mp

-13--10 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1[N+]([O-])=O

InChI

1S/C8H9NO2/c1-2-7-5-3-4-6-8(7)9(10)11/h3-6H,2H2,1H3

InChI key

PXWYZLWEKCMTEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-2-nitrobenzene may be used as a solvent to compose the supported liquid membrane (SLM) for use in electromembrane extractions (EMEs).[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Šlampová et al.
Electrophoresis, 35(23), 3317-3320 (2014-09-17)
Selectivity of electromembrane extractions (EMEs) was fine-tuned by modifications of supported liquid membrane (SLM) composition using additions of various 18-crown-6 ethers into 1-ethyl-2-nitrobenzene. Gradually increased transfer of K(+) , the cation that perfectly fits the cavity of 18-crown-6 ethers, was
Chuixiu Huang et al.
Journal of chromatography. A, 1326, 7-12 (2014-01-01)
In this work, a single-well electromembrane extraction (EME) device was developed based on a thin (100μm) and flat porous membrane of polypropylene supporting a liquid membrane. The new EME device was operated with a relatively large acceptor solution volume to
Andrea Šlampová et al.
Analytica chimica acta, 887, 92-100 (2015-09-01)
Electrolysis is omnipresent in all electrochemical processes including electromembrane extraction (EME). The effects of electrolysis on quantitative aspects of EME were comprehensively evaluated for a set of acidic (substituted phenols) and basic (basic drugs) analytes. EMEs were carried out across

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service