Skip to Content
Merck
All Photos(1)

Key Documents

536393

Sigma-Aldrich

4,4′-Dinitrodiphenyl ether

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$70.90

$70.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$70.90

About This Item

Linear Formula:
(NO2C6H4)2O
CAS Number:
Molecular Weight:
260.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$70.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

140-145 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(Oc2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C12H8N2O5/c15-13(16)9-1-5-11(6-2-9)19-12-7-3-10(4-8-12)14(17)18/h1-8H

InChI key

MWAGUKZCDDRDCS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Seiler
Carcinogenesis, 6(12), 1811-1813 (1985-12-01)
The results of Salmonella mutagenicity tests with the herbicide nitrofen (2,4-dichlorophenyl-4-nitrophenyl ether, CAS 1836-75-5) reported in the literature seem to vary according to source or lot of the technical product. This behaviour could be reproduced in our experiments, and it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service