Skip to Content
Merck
All Photos(1)

Key Documents

536369

Sigma-Aldrich

2,6-Dichlorodiphenylamine

98%

Synonym(s):

N-Phenyl-2,6-dichloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHC6H5
CAS Number:
Molecular Weight:
238.11
Beilstein:
2725987
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

50-53 °C (lit.)

SMILES string

Clc1cccc(Cl)c1Nc2ccccc2

InChI

1S/C12H9Cl2N/c13-10-7-4-8-11(14)12(10)15-9-5-2-1-3-6-9/h1-8,15H

InChI key

HDUUZPLYVVQTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service