Skip to Content
Merck
All Photos(1)

Key Documents

536008

Sigma-Aldrich

5-(Aminomethyl)-2-chloropyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$219.00
25 G
$440.00

$219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$219.00
25 G
$440.00

About This Item

Empirical Formula (Hill Notation):
C6H7ClN2
CAS Number:
Molecular Weight:
142.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$219.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

28-34 °C

functional group

amine
chloro

SMILES string

NCc1ccc(Cl)nc1

InChI

1S/C6H7ClN2/c7-6-2-1-5(3-8)4-9-6/h1-2,4H,3,8H2

InChI key

XPARFBOWIYMLMY-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret L Eng et al.
The Science of the total environment, 760, 143409-143409 (2020-11-22)
Neonicotinoids are the most widely used insecticides globally, but their rapid metabolism in vertebrates makes diagnosing wildlife exposure challenging. More detailed information on the pattern of imidacloprid metabolites over time could be used to better approximate the timing and level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service