Skip to Content
Merck
All Photos(1)

Key Documents

535559

Sigma-Aldrich

Ethylene carbonate

anhydrous, 99%

Synonym(s):

1,3-Dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
Beilstein:
106249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

anhydrous

vapor density

3.04 (vs air)

vapor pressure

0.02 mmHg ( 36.4 °C)

Assay

99%

impurities

<0.005% water

bp

243-244 °C/740 mmHg (lit.)

mp

35-38 °C (lit.)

density

1.321 g/mL at 25 °C (lit.)

SMILES string

O=C1OCCO1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang-Jae Park et al.
Journal of the American Chemical Society, 137(7), 2565-2571 (2015-02-04)
Here we describe a class of electric-conducting polymers that conduct electrons via the side chain π-π stacking. These polymers can be designed and synthesized with different chemical moieties to perform different functions, extremely suitable as a conductive polymer binder for
S Ramesh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(4-5), 1237-1242 (2006-08-22)
The polymer electrolytes composing of the blend of polyvinyl chloride-polymethyl methacrylate (PVC/PMMA) with lithium triflate (LiCF3SO3) as salt, ethylene carbonate (EC) and dibutyl phthalate (DBP) as plasticizers and silica (SiO2) as the composite filler were prepared. FTIR studies confirm the
T R Hanley et al.
Toxicology and applied pharmacology, 100(1), 24-31 (1989-08-01)
Ethylene carbonate (EC) has a toxicity profile which resembles that of ethylene glycol (EG). To determine whether the toxicity of EC could be explained on the basis of its metabolism to EG, male Fischer 344 rats were given 200 mg/kg
Y L Yang et al.
The Journal of biological chemistry, 273(14), 7814-7817 (1998-05-09)
Ethylene carbonate, a cyclic organic carbonate widely used industrially, is toxic when metabolically converted to ethylene glycol. A rat liver enzyme active in catalyzing the ring opening has been purified to electrophoretic homogeneity and found to be active in the
Niccolò Peruzzi et al.
The journal of physical chemistry. B, 116(49), 14398-14405 (2012-11-24)
The solubility of some potassium salts (KF, KCl, KBr, KI, KNO(3), KClO(4), KSCN, and KSeCN) in ethylene carbonate (EC) was determined at different temperatures with an inductively coupled plasma atomic emission spectrometer. From the solubility measurements, the thermodynamic parameters ΔG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service