Skip to Content
Merck
All Photos(2)

Documents

533963

Sigma-Aldrich

Poly[[6-[(1,1,3,3-tetramethylbutyl)amino]-s-triazine-2,4-diyl]-[(2,2,6,6-tetramethyl-4-piperidyl)imino]-hexamethylene-[(2,2,6,6-tetramethyl-4-piperidyl)imino]

average Mn ~2,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C35H68N8)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn ~2,000

mp

136-140 °C (lit.)

λmax

240 nm

InChI

1S/C24H50N4.C8H19N.C3Cl3N3/c1-21(2)15-19(16-22(3,4)27-21)25-13-11-9-10-12-14-26-20-17-23(5,6)28-24(7,8)18-20;1-7(2,3)6-8(4,5)9;4-1-7-2(5)9-3(6)8-1/h19-20,25-28H,9-18H2,1-8H3;6,9H2,1-5H3;

InChI key

ORECYURYFJYPKY-UHFFFAOYSA-N

Related Categories

General description

Poly[[6-[(1,1,3,3-tetramethylbutyl)amino]-s-triazine-2,4-diyl]-[(2,2,6,6-tetramethyl-4-piperidyl)imino]-hexamethylene-[(2,2,6,6-tetramethyl-4-piperidyl)imino](HAS) is an amine hindered light stabilizer that can be used as an antioxidant by attaching the nitroxyl radical with the surface atoms.

Application

Hindered amine UV light stabilizer for polyolefins.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterization of Hindered Amine Light Stabilizers in Polymer Matrix Using Terahertz Time-Domain Spectroscopy
Xie L, et al.
Macromolecular Chemistry and Physics, 213(14), 1441-1447 (2012)
UPLC-MS as a powerful technique for screening the nonvolatile contaminants in recycled PET
Bentayeb K, et al.
Analytical and Bioanalytical Chemistry, 388(5-6), 1031-1038 (2007)
EPR study of the formation of radicals in PP with antioxidants irradiated with gamma rays
Silva P, et al.
Nuclear Instruments & Methods in Physics Research. Section B, Beam Interactions With Materials and Atoms, 265(1), 300-304 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service