Skip to Content
Merck
All Photos(1)

Documents

532665

Sigma-Aldrich

(4aR)-trans-5-(1,5,5,8aS-Tetramethyl-2-methylenedecahydro-1-naphthalenyl)-(3R)-methyl-1-penten-3-ol

98%

Synonym(s):

Manool

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O
CAS Number:
Molecular Weight:
290.48
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +27°, c = 1 in chloroform

mp

49-52 °C (lit.)

SMILES string

CC1(C)CCC[C@]2(C)[C@@H](CC[C@@](C)(O)C=C)C(=C)CC[C@@H]12

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service