Skip to Content
Merck
All Photos(1)

Key Documents

53096

Sigma-Aldrich

1-Ethyl-3-methylimidazolium chloride

dry, ≥98.0% (T)

Synonym(s):

1-Methyl-3-ethylimidazolinium chloride, 1-Methyl-3-ethylimidazolium chloride, 3-Ethyl-1-methylimidazolium chloride, N-Methyl-N′-ethylimidazolium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$103.00
25 G
$384.00

$103.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$103.00
25 G
$384.00

About This Item

Empirical Formula (Hill Notation):
C6H11ClN2
CAS Number:
Molecular Weight:
146.62
Beilstein:
5163190
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (T)

form

solid

impurities

≤800 ppm water

mp

77-79 °C (lit.)

SMILES string

[Cl-].CCn1cc[n+](C)c1

InChI

1S/C6H11N2.ClH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1

InChI key

BMQZYMYBQZGEEY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anshu Sharma et al.
Carbohydrate polymers, 134, 617-626 (2015-10-03)
Ionic liquids (IL) can alter the physical properties of agar hydrogels. Rheology studies show that gels with wide range of storage moduli (gel strength) G0 values ranging from 1 to 20 KPa could be made in imidazolium based IL solutions
M T Ubeda-Torres et al.
Journal of chromatography. A, 1380, 96-103 (2015-01-17)
In reversed-phase liquid chromatography in the absence of additives, cationic basic compounds give rise to broad and asymmetrical peaks as a result of ionic interactions with residual free silanols on silica-based stationary phases. Ionic liquids (ILs), added to the mobile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service