Skip to Content
Merck
All Photos(1)

Key Documents

52917

Sigma-Aldrich

1,4,7,10,13,16-Hexathiacyclooctadecane

≥97.0% (S)

Synonym(s):

18-Ane-S6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24S6
CAS Number:
Molecular Weight:
360.71
Beilstein:
143957
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

Assay

≥97.0% (S)

mp

92-94 °C (lit.)
92-96 °C

SMILES string

C1CSCCSCCSCCSCCSCCS1

InChI

1S/C12H24S6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2

InChI key

HELCSRZMDRYPIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Crown-compound forming complexes with transition metal ions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.E. Wolf et al.
Inorg. Synth., 25, 122-122 (1989)
E. Sekido et al.
Bull. Chem. Soc. Bull., 61, 1587-1587 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service