Skip to Content
Merck
All Photos(1)

Key Documents

528986

Sigma-Aldrich

2-Fluoro-4-methoxybenzaldehyde

97%

Synonym(s):

2-Fluoro-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
154.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

43-48 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(C=O)c(F)c1

InChI

1S/C8H7FO2/c1-11-7-3-2-6(5-10)8(9)4-7/h2-5H,1H3

InChI key

UNWQNFJBBWXFBG-UHFFFAOYSA-N

General description

2-Fluoro-4-methoxybenzaldehyde is a fluorinated aromatic aldehyde. It can be prepared from 4-bromo-3-fluoroanisole.[1]

Application

2-Fluoro-4-methoxybenzaldehyde may be used in the preparation of:
  • fluorine containing 2,4,5-trisubstituted imidazole[2]
  • 1-(2-fluoro-4-methoxyphenyl)-2-propanone[3]
  • 6-(2-fluoro-4-methoxyphenyl)fulvene[4]
  • 10-(2-fluoro-4-methoxyphenyl)-6,7,9,10-tetrahydro-1Hfuro[3,4-b]pyrazolo[3,4-f]quinolin-9-one[5]
  • polyhydroquinoline (PHQ)[6]
  • 3-(2-fluoro-4-methoxyphenyl) acrylic acid methyl ester[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indium trifluoride: A highly efficient catalyst for the synthesis of fluorine-containing 2, 4, 5-trisubstituted imidazoles under solvent-free conditions.
Reddy MV and Jeong YT.
Journal of Fluorine Chemistry, 142, 45-51 (2012)
Synthesis of 1, 3-Bis (hydroxy-halogenophenyl)-propane-1, 3-diamines and their Pt (II) Complexes, Syntheses of the Ligands.
Kammermeier T and Wiegrebe W.
Arch. Pharm. (Weinheim), 327, 547-561 (1994)
Fluorinated derivatives of titanocene Y: synthesis and cytotoxicity studies.
Claffey J, et al.
European Journal of Organic Chemistry, 26, 4074-4082 (2008)
Polystyrene-Supported p-Toluenesulfonic Acid: A New, Highly Efficient, and Recyclable Catalyst for the Synthesis of Hydropyridine Derivatives under Solvent-Free Conditions.
Reddy MV and Jeong YT.
Synlett, 23(20), 2985-2991 (2012)
Synthesis, cytotoxic activity and docking studies of new 4-aza-podophyllotoxin derivatives.
Hatti I, et al.
Medicinal Chemistry Research, 24(8), 3305-3313 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service