Skip to Content
Merck
All Photos(1)

Key Documents

528862

Sigma-Aldrich

2-Bromo-4,6-dimethylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(CH3)2NH2
CAS Number:
Molecular Weight:
200.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

43-47 °C (lit.)

functional group

bromo

SMILES string

Cc1cc(C)c(N)c(Br)c1

InChI

1S/C8H10BrN/c1-5-3-6(2)8(10)7(9)4-5/h3-4H,10H2,1-2H3

InChI key

YOSJCQJJIHEUKA-UHFFFAOYSA-N

Application

2-Bromo-4,6-dimethylaniline may be used in the synthesis of 2-phenoxy-4,6-dimethylaniline[1] and (4aR,9aR)-4a,6,8-trimethyl-4,4a,9,9a-tetrahydro-1H-carbazol-2(3H)-one.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using intelligent/random library screening to design focused libraries for the optimization of homogeneous catalysts: Ullmann ether formation.
Fagan PJ, et al.
Journal of the American Chemical Society, 122(21), 5043-5051 (2000)
Palladium-catalyzed gamma-arylation of beta,gamma-unsaturated ketones: application to a one-pot synthesis of tricyclic indolines.
Alan M Hyde et al.
Angewandte Chemie (International ed. in English), 47(1), 177-180 (2007-11-23)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service