All Photos(1)
About This Item
Linear Formula:
C2H5OC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
134-138 °C (lit.)
SMILES string
CCOc1ccccc1\C=C\C(O)=O
InChI
1S/C11H12O3/c1-2-14-10-6-4-3-5-9(10)7-8-11(12)13/h3-8H,2H2,1H3,(H,12,13)/b8-7+
InChI key
UXTDCJJEJZCEBF-BQYQJAHWSA-N
General description
2-Ethoxycinnamic acid is a cinnamic acid derivative. The aromatic and alkene group in this molecule are coplanar.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Methyl (Z)-2-chloro-3-(2-methoxycarbonylphenyl) prop-2-enoate.
Fadgen CJ, et al.
Acta Crystallographica Section E, Structure Reports Online, 66(2), o376-o376 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service