Skip to Content
Merck
All Photos(1)

Key Documents

528226

Sigma-Aldrich

5-Bromo-2(1H)-pyridone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$88.40
25 G
$110.00

$88.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$88.40
25 G
$110.00

About This Item

Empirical Formula (Hill Notation):
C5H4BrNO
CAS Number:
Molecular Weight:
174.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

180-183 °C (lit.)

functional group

bromo

SMILES string

BrC1=CNC(=O)C=C1

InChI

1S/C5H4BrNO/c6-4-1-2-5(8)7-3-4/h1-3H,(H,7,8)

InChI key

NDMZZQRNZFWMEZ-UHFFFAOYSA-N

General description

5-Bromo-2(1H)-pyridone undergoes difluormethylation in the presence of sodium chlorodifluoroacetate (ClCF2COONa) and methyl cyanide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makoto Ando et al.
Organic letters, 8(17), 3805-3808 (2006-08-11)
[reaction: see text] A novel one-pot synthesis of N-difluoromethyl-2-pyridones is described. N-(Pyridin-2-yl)acetamide derivatives were excellent precursors for the preparation of N-difluoromethyl-2-pyridone derivatives. Difluoromethylation of 2-acetaminopyridine derivatives was achieved with sodium chlorodifluoroacetate as a difluorocarbene source in the presence of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service