Skip to Content
Merck
All Photos(1)

Key Documents

524794

Sigma-Aldrich

3-Aminophthalimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

268.5-273.5 °C (lit.)

SMILES string

Nc1cccc2C(=O)NC(=O)c12

InChI

1S/C8H6N2O2/c9-5-3-1-2-4-6(5)8(12)10-7(4)11/h1-3H,9H2,(H,10,11,12)

InChI key

GQBONCZDJQXPLV-UHFFFAOYSA-N

General description

3-Aminophthalimide (API) is a stable precursor of luminol.[1] API can be prepared by the hydrogenation of 3-nitrophthalimide and methanol in the presence of 10% Pd-C.[2]

Application

3-Aminophthalimide may be used in direct chemiluminescent cellular bioassays.[1] It may also be used in the preparation of 3-bromopthalimide.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Study of the Aminophthalimidoacetic Acids1.
Caswell LR and Atkinson PC.
The Journal of Organic Chemistry, 29(11), 3151-3154 (1964)
Demetra Mavri-Damelin et al.
Bioconjugate chemistry, 20(2), 266-273 (2009-01-15)
We present a labeling system for direct chemiluminescence-based cellular bioassays using the stable pro-chemiluminescent, luminol precursor, 3-aminophthalimide (API). API-coupled reporter molecules are detected chemiluminometrically after treatment with hydrazine, which converts the API label to luminol. API derivatives containing a variety
177. Chemiluminescent organic compounds. Part VII. Substituted phthalaz-1: 4-diones. Effect of substituents on the luminescent power.
Drew HDK and Garwood RF.
Journal of the Chemical Society, 836-837 (1939)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service