Skip to Content
Merck
All Photos(1)

Key Documents

524719

Sigma-Aldrich

Phenylzinc bromide solution

0.5 M in THF

Synonym(s):

Bromo(phenyl)zinc, Bromophenylzinc, Phenylzinc bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$265.00

$265.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$265.00

About This Item

Linear Formula:
C6H5ZnBr
CAS Number:
Molecular Weight:
222.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$265.00


Check Cart for Availability

Request a Bulk Order

concentration

0.5 M in THF

bp

65 °C

density

0.99 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]c1ccccc1

InChI

1S/C6H5.BrH.Zn/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1

InChI key

LMUFDZHWZIRZOY-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service