Skip to Content
Merck
All Photos(1)

Key Documents

523038

Sigma-Aldrich

Platinum(II) acetylacetonate

≥99.98% trace metals basis

Synonym(s):

2,4-Pentanedione platinum(II) derivative, Pt(acac)2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$777.00

$777.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$777.00

About This Item

Linear Formula:
Pt(C5H7O2)2
CAS Number:
Molecular Weight:
393.29
Beilstein:
4136189
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$777.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.98% trace metals basis

form

solid

reaction suitability

core: platinum

mp

249-252 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Pt]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pt/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

KLFRPGNCEJNEKU-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Platinum(II) acetylacetonate is high-purity compound that is widely used as a key precursor in chemical vapor deposition (CVD) and atomic layer deposition (ALD) methods, to facilitate the synthesis of platinum thin films. These films are utilized in microelectronics, sensors, and catalytic coatings due to platinum′s high thermal stability, chemical inertness, and catalytic activity. Additionally, Pt(acac)₂ is used to synthesize platinum nanoparticles, essential for applications in catalysis and fuel cells.

Application

Platinum(II) acetylacetonate (Pt(acac)₂) is used as a precursor in the synthesis of:
  • Pt−Co octahedral nanocrystals, which are used to improve catalytic activity and durability in the oxygen reduction reaction (ORR) in fuel cells.
  • PtW@WO3 NPs via thermal decomposition method, serving as efficient catalysts for the hydrogen evolution reaction (HER).
  • Monodisperse Pt–Fe nanoparticles, which exhibit high durability for proton exchange membrane fuel cells and provide active and durable low-Pt electrocatalysts for electrochemical devices.

Features and Benefits

Platinum(II) acetylacetonate exhibits
  • High Purity: Reduces contamination risk, ensuring reliable performance in applications.
  • Thermal Stability: Maintains activity at elevated temperatures, enhancing product lifespan and efficiency.
  • Soluble in acetone
  • Elemental Carbon is >29.5% and Platinum presents >47.9%

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service