Skip to Content
Merck
All Photos(2)

Key Documents

520667

Sigma-Aldrich

Palladium on alumina

greener alternative

extent of labeling: 0.5 wt. % loading, dry, tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

dry
tablet

reaction suitability

reagent type: catalyst
core: palladium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

0.5 wt. % loading

mp

1552 °C (lit.)

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palladium on alumina (Pd/Al2O3) (0.5 wt % loading, dry, tablet) is a widely utilized catalyst that can be prepared by wet impregnation of alumina pellets with palladium chloride.[1] It can also be prepared by flame spray pyrolysis (FSP) by utilizing the precursor solution of aluminum sec-butoxide and palladium acetylacetonate.[2]

Application

Pd/Al2O3 can be used as a catalyst in the enantioselective hydrogenation reaction.[2] It can also be potentially used in the formation of an automotive catalytic converter to improve the air quality of urban environments.[3][4]

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic activity of Pd/Al2O3 toward the combustion of methane
Gao D, et al.
Catalysis Communications, 9(15), 2583-2587 (2008)
Flame spray synthesis of Pd/Al2O3 catalysts and their behavior in enantioselective hydrogenation
Strobel R, et al.
J. Catal., 222(2), 307-314 (2004)
Aqueous bromate reduction by catalytic hydrogenation over Pd/Al2O3 catalysts
Chen H, et al.
Applied Catalysis. B, Environmental, 96(3-4), 307-313 (2010)
Identification of the phytosphingosine metabolic pathway leading to odd-numbered fatty acids
Kondo N, et al.
Nature Communications, 5(1), 1-13 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service