Skip to Content
Merck
All Photos(1)

Key Documents

520543

Sigma-Aldrich

Pyridine-d5 N-oxide

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$167.00

$167.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
1 G
$167.00

About This Item

Empirical Formula (Hill Notation):
C5D5NO
CAS Number:
Molecular Weight:
100.13
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

$167.00


Estimated to ship on29 May 2025


Request more information

isotopic purity

98 atom % D

bp

270 °C (lit.)

mp

61-65 °C (lit.)

mass shift

M+5

SMILES string

[O-][N+]1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]

InChI

1S/C5H5NO/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

ILVXOBCQQYKLDS-RALIUCGRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service